Learn More
SYNGR4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SYNGR4.
Brand: Abnova PAB6719.100ug
Description
This gene encodes an integral membrane protein. The gene belongs to the synaptogyrin gene family. Like other members of the family the protein contains four transmembrane regions. The exact function of this protein is unclear. [provided by RefSeq
Sequence: HIPKSLQELANS-CSpecifications
| SYNGR4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.01-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SYNGR4 | |
| SYNGR4 | |
| A synthetic peptide corresponding to human SYNGR4. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SYNGR4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC125805 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 23546 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only