Learn More
STAB1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of STAB1.
Brand: Abnova PAB18612.100ug
Description
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node. The receptor has been shown to endocytose ligands such as low density lipoprotein, Gram-positive and Gram-negative bacteria, and advanced glycosylation end products. Supporting its possible role as a scavenger receptor, the protein rapidly cycles between the plasma membrane and early endosomes. [provided by RefSeq
Sequence: C-DFPDTQRILTVKSpecifications
| STAB1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(3-5 ug/mL) ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
| STAB1 | |
| STAB1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human STAB1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of STAB1. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| CLEVER-1/FEEL-1/FELE-1/FEX1/KIAA0246/STAB-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 23166 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only