Learn More
SOX1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SOX1.
Brand: Abnova PAB7441.100ug
Description
This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. In mice, a similar protein regulates the gamma-crystallin genes and is essential for lens development. [provided by RefSeq
Sequence: C-AHPHNPQPMHRSpecifications
| SOX1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) The optimal working dilution should be determined by the end user. | |
| SOX1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 6656 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SOX1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| SOX1 | |
| A synthetic peptide corresponding to human SOX1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only