Learn More
SNX15, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SNX15.
Brand: Abnova PAB6367.100ug
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: C-EEILRLHLSQLPPSpecifications
| SNX15 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SNX15 | |
| SNX15 | |
| A synthetic peptide corresponding to human SNX15. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both human isoforms according to NP_037438.2 and NP_680086.2. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SNX15. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HSAF001435 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 29907 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only