Learn More
SLC17A5, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SLC17A5.
Brand: Abnova PAB27673.100ug
Description
This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form. [provided by RefSeq
Sequence: EHSAPIKVHHNQTGKSpecifications
| SLC17A5 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user. | |
| SLC17A5 | |
| SLC17A5 | |
| A synthetic peptide corresponding to amino acids 85-99 at internal region of human SLC17A5. | |
| 100 μg | |
| Primary | |
| Human |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SLC17A5. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| AST/FLJ22227/FLJ23268/ISSD/NSD/SD/SIALIN/SIASD/SLD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 26503 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only