Learn More
SDCBP, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SDCBP.
Brand: Abnova PAB7132.100ug
Description
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: SLEDLKVDKVIQAQ-CSpecifications
| SDCBP | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| SDCBP | |
| SDCBP | |
| A synthetic peptide corresponding to human SDCBP. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_005616.2 and NP_001007068.1) and isoform 3 (NP_001007070.1and NP_001007071.1) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SDCBP. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MDA-9/ST1/SYCL/TACIP18 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 6386 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only