missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUFY1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RUFY1.
Brand: Abnova PAB18985.100ug
This item is not returnable.
View return policy
Description
Sequence: C-QKNEAITSFEGKTSpecifications
| RUFY1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.05-0.2 ug/mL) The optimal working dilution should be determined by the end user. | |
| RUFY1 | |
| RUFY1 | |
| A synthetic peptide corresponding to amino acids at internal region of human RUFY1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_079434.3; NP_001035541.1). Reported variants represent identical protein: NP_001035542.1, NP_001035541.1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of RUFY1. | |
| In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FLJ22251/RABIP4/ZFYVE12 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 80230 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction