Learn More
RGS14, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RGS14.
Brand: Abnova PAB6630.100ug
Description
This gene encodes a member of the regulator of G-protein signaling family. This protein contains one RGS domain, two Raf-like Ras-binding domains (RBDs), and one GoLoco domain. The protein attenuates the signaling activity of G-proteins by binding, through its GoLoco domain, to specific types of activated, GTP-bound G alpha subunits. Acting as a GTPase activating protein (GAP), the protein increases the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Sequence: C-IGGSLNSTTDSALSpecifications
| RGS14 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| RGS14 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10636 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of RGS14. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| RGS14 | |
| A synthetic peptide corresponding to human RGS14. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only