missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL2/RFPL3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RFPL2/RFPL3.
Brand: Abnova PAB6329.100ug
This item is not returnable.
View return policy
Description
Sequence: C-TTDAPVRPGEAKSpecifications
| RFPL2/RFPL3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| RNF79 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10739/10738 | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of RFPL2/RFPL3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| RFPL2 | |
| A synthetic peptide corresponding to human RFPL2/RFPL3. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both RFPL2 (NP_006596) and RFPL3 (NP_006595), which are virtually identical. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction