missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgM Fc Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69746
371.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Rat samples. It is validated for Western Blot, ELISA, Dot Blot
Specifications
| IgM Fc | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Secondary | |
| Rat | |
| Lyophilized |
| Western Blot, ELISA, Dot Blot | |
| LYOPH | |
| Lyophilized from PBS (pH 7.2) | |
| Purified homogenous IgM isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 10 mg | |
| B Cell Development and Differentiation Markers, Immunology | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction