missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2c Heavy Chain Secondary Antibody, DyLight 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7132UV
This item is not returnable.
View return policy
Description
IgG2c Heavy Chain Polyclonal antibody specifically detects IgG2c Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2c Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rat IgG2c | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction