missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2b Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB7127
This item is not returnable.
View return policy
Description
IgG2b Heavy Chain Polyclonal antibody specifically detects IgG2b Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgG2b Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| By immunoelectrophoresis and ELISA this reacts specifically with Rat IgG2b. This may cross react with IgG2b from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000 (Primary) 1:100-1:500 (For Coating Plates), Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin | |
| Rat IgG2b | |
| 1 mL | |
| Secondary | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction