missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2a Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB7122
This item is not returnable.
View return policy
Description
IgG2a Heavy Chain Polyclonal antibody specifically detects IgG2a Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgG2a Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| By immunoelectrophoresis and ELISA this reacts specifically with Rat IgG2a. This may cross react with IgG2a from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000 (Primary) 1:100-1:500 (for coating plates), Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000 | |
| Rat IgG2a | |
| 1 mL | |
| Secondary | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction