missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rat IgG (H+L) Secondary Antibody, FITC

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen™ 31629


Product Code. 11899180

  • £151.00 / 2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31629 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31629 reacts with the heavy chains of rat IgG and with light chains common to most rat immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, antibodies may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 10.2 μg/mg; 3.8 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rat IgG (H+L)
Polyclonal
FITC
Goat
2 mg
Secondary
4°C, store in dark
Lyophilized
Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
1.5 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Antigen affinity chromatography
RUO
Rat
Secondary Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Goat anti-Rat IgG (H+L) Secondary Antibody, FITC

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : SODIUM AZIDE

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.