missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Rat IgG (H+L) Secondary Antibody
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ SA510278
This item is not returnable.
View return policy
Description
This antibody reacts with rat IgG and with light chains common with other rat immunoglobulins as determined by IEP and ELISA techniques. This antibody may cross react with IgG from other species.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rat IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Rat IgG h+l. | |
| 1 mg | |
| Secondary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction