Learn More
Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen SA510019
154.21 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : PN
Additional Details : Weight : 0.01000kg
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, human, mouse, rabbit and sheep.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 550 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Rat IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |