missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rat IgG Fc Secondary Antibody, DyLight 650, Novus Biologicals™

Product Code. 18423126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423126 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423126 Supplier Novus Biologicals Supplier No. NBP262582C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG Fc Polyclonal antibody specifically detects IgG Fc in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Rat IgG F(c) fragment
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.