missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IgE Fc Polyclonal antibody specifically detects IgE Fc in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
Specifications
Specifications
| Antigen | IgE Fc |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting, Dot Blot |
| Formulation | Lyophilized from PBS (pH 7.2) |
| Host Species | Goat |
| Immunogen | Pools of purified homogenous IgE isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure |
| Purification Method | Ion exchange chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?