missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgA Fc Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69215
This item is not returnable.
View return policy
Description
IgA Fc Polyclonal antibody specifically detects IgA Fc in Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
Specifications
| IgA Fc | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.2) | |
| Purified homogenous IgA isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 10 mg | |
| Secondary | |
| Rat | |
| Lyophilized |
| Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
| LYOPH | |
| Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction