Learn More
Invitrogen™ Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, Biotin
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Specifications
Specifications
| Antigen | Rabbit IgG (Heavy chain) |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Superclonal |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide |
| Host Species | Goat |
| Immunogen | Recombinant full-length protein. |
| Purification Method | Gravity column chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.