missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6719
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500-1:2500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin 1:50 - 1:200, Dot Blot, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Rabbit IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Rabbit IgG (H and L) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, ELISA | |
| Texas Red | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction