Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ 656122
Product Code. 10406413
  • £186.00 / 1mL
For Research Use Only. All usage must comply with product instructions.
Description
Description
Goat anti-Rabbit IgG (H+L) Secondary Antibody for ELISA, IHC (P), IP, Western Blot
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit IgG (H+L)
Polyclonal
Alkaline Phosphatase (AP)
Goat
Affinity chromatography
RUO
Rabbit
Secondary Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
0.05M tris with 0.15M NaCl, 1mM MgCl2, 1% BSA, 50% glycerol and 0.1% sodium azide; pH 7.5
Gamma Immunoglobin.
1 mL
Secondary
4°C