missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG - H&L (HRP), Secondary Antibody, Abcam™
Description
Host Species: Goat
Isotype: IgG
Applications:
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | Rabbit IgG - H&L (HRP) |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | HRP |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Research Discipline | Immunology, Secondary Antibody |
| Primary or Secondary | Secondary |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?