missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, PE-Alexa Fluor™ 610
Description
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody for Flow
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | PE-Alexa Fluor 610 |
| Formulation | PBS with 2mM EDTA, 1% glycerol and 5mM sodium azide; pH 7.5 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Purification Method | Purified |
| Quantity | 100 μg |
| Show More |
Frequently Asked Questions (FAQs)
Can I use my tandem dye antibody (e.g., Alexa Fluor 610-R-phycoerythrin) for microscopy?
Tandem dyes are not recommended for imaging applications. Both R-PE or APC tend to photobleach quickly and cannot be archived. Any unfolding of the phycobiliprotein would result in loss of fluorescence. They work great in flow cytometry applications.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?