missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Goat Polyclonal Secondary Antibody

£79.75 - £205.00

Specifications

Content And Storage Store at 2–8°C. For long term storage of antibody solutions, dilute with 50% glycerol, aliquot, and store at -5 to -35°C.
Applications Immunoassay, Immunohistochemistry, Western Blot
Conjugate HRP
Form Lyophilized
Host Species Goat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15393916
View Documents
Invitrogen™
A16104SAMPLE
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15281388
View Documents
Invitrogen™
A16104
1 mg
£205.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rabbit serum proteins, serum proteins from bovine, human, or mouse , IgG from human or mouse. Based on IEP, this antibody reacts with: heavy (gamma) chains on rabbit IgG; light chains on all rabbit immunoglobulins. Affinity purified antibody is > 95% based on SDS-PAGE.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Store at 2–8°C. For long term storage of antibody solutions, dilute with 50% glycerol, aliquot, and store at -5 to -35°C.
HRP
Goat
RUO
Lyophilized
Affinity Purified
Immunoassay, Immunohistochemistry, Western Blot
Lyophilized
IgG
Mouse
Secondary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.