Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 350
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 62272
Description
Product # 62272 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 62272 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, horse, mouse, swine, goat, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 350 Amax= 353 nm; Emax= 432 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| DyLight 350 | |
| Goat | |
| 500 μL | |
| Secondary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| Antigen affinity chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.