Learn More
Invitrogen™ Goat anti-Rabbit IgG Fc Secondary Antibody
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31216 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31216 reacts with the Fc portion of rabbit IgG heavy chain but not with the Fab portion of rabbit immunoglobulins. The antibody does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Specifications
Specifications
| Antigen | Rabbit IgG Fc |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 2 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.