Learn More
Invitrogen™ Goat anti-Rabbit IgG F(ab')2 Secondary Antibody, Biotin
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 31823
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31823 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31823 reacts with the light chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against the Fc portion of rabbit IgG or against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 2.0 mL of distilled water. Country of Origin: USA.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG F(ab')2 | |
| Polyclonal | |
| Biotin | |
| Goat | |
| 2 mL | |
| Secondary | |
| 4°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.6 mg/mL | |
| PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
| Affinity chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.