Learn More
PRDX1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PRDX1.
Brand: Abnova PAB11441.100ug
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]
Sequence: C-SDPKRTIAQDYGSpecifications
| PRDX1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRDX1 | |
| PRDX1 | |
| A synthetic peptide corresponding to human PRDX1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PRDX1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MSP23/NKEFA/PAG/PAGA/PAGB/PRX1/PRXI/TDPX2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5052 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only