Learn More
PRAM1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PRAM1.
Brand: Abnova PAB6052.100ug
Description
The protein encoded by this gene is similar to FYN binding protein (FYB/SLAP-130), an adaptor protein involved in T cell receptor mediated signaling. This gene is expressed and regulated during normal myelopoiesis. The expression of this gene is induced by retinoic acid and is inhibited by the expression of PML-RARalpha, a fusion protein of promyelocytic leukemia (PML) and the retinoic acid receptor-alpha (RARalpha). [provided by RefSeq
Sequence: DFCDPLENQPLPLGRSpecifications
| PRAM1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRAM1 | |
| PRAM1 | |
| A synthetic peptide corresponding to human PRAM1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PRAM1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC39864/PRAM-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84106 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only