Learn More
PPP2R1A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PPP2R1A.
Brand: Abnova PAB6772.100ug
Description
This gene encodes a constant regulatory subunit of protein phosphatase 2. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. This gene encodes an alpha isoform of the constant regulatory subunit A. [provided by RefSeq
Sequence: C-KYFAQEALTVLSLASpecifications
| PPP2R1A | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PPP2R1A | |
| PPP2R1A | |
| A synthetic peptide corresponding to human PPP2R1A. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PPP2R1A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC786/PR65A | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5518 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only