missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Porcine IgM Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-36559
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Phosphate Buffered Saline (PBS) with 0.09% Sodium Azide | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Pig IgM | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with pig IgM. This antibody may cross react with IgM from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction