missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Porcine IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB738
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000(Primary) 1:100-1:500(for coating plates), Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000, Immunohistochemistry-Paraffin 1:200-1:2000 | |
| Pig IgG-heavy and light chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| By immunoelectrophoresis and ELISA this reacts specifically with pig IgG and with light chains common to other pig immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction