missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHB1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PLEKHB1.
Brand: Abnova PAB19683.100ug
This item is not returnable.
View return policy
Description
Sequence: PPDSALESPFEE-CSpecifications
| PLEKHB1 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of PLEKHB1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| KPL1/PHR1/PHRET1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 58473 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PLEKHB1 | |
| PLEKHB1 | |
| A synthetic peptide corresponding to N-terminus of human PLEKHB1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize reported isoforms a (NP_067023.1) and c (NP_001123505.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction