Learn More
PIR, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PIR.
Brand: Abnova PAB6662.100ug
Description
This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described. [provided by RefSeq
Sequence: GSSKKVTLSVLSRECSpecifications
| PIR | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PIR | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8544 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PIR. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PIR | |
| A synthetic peptide corresponding to human PIR. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only