missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Pig IgG (H+L) Secondary Antibody, FITC
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA184850
This item is not returnable.
View return policy
Description
Goat anti Pig IgG (H/L) antibody recognizes porcine IgG (heavy and light chains) and has been shown to react specifically with porcine IgG and with light chains common to other pig immunoglobulins, by immunoelectrophoresis and ELISA.
Anti-Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for porcine (swine, pig) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Pig IgG (H+L) | |
| Polyclonal | |
| FITC | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Pig | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Proprietary Immunogen. | |
| 1 mg | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction