Learn More
PGBD2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PGBD2.
Brand: Abnova PAB7425.100ug
Description
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. The exact function of this gene is not known. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-ESNADTTSQGRRSRRSpecifications
| PGBD2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| PGBD2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 267002 | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PGBD2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PGBD2 | |
| A synthetic peptide corresponding to human PGBD2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001017434.1, NP_733843.1) | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only