Learn More
PDK1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PDK1.
Brand: Abnova PAB6898.100ug
Description
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. [provided by RefSeq
Sequence: C-DFKDKSAEDAKSpecifications
| PDK1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PDK1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5163 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PDK1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PDK1 | |
| A synthetic peptide corresponding to human PDK1. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only