Learn More
PCOLCE, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PCOLCE.
Brand: Abnova PAB27535.100ug
Description
Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity. [provided by RefSeq
Sequence: QILTNLSKRKCPSQPSpecifications
| PCOLCE | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.3-1ug/ml) The optimal working dilution should be determined by the end user. | |
| PCOLCE | |
| PCOLCE | |
| A synthetic peptide corresponding to C-terminus of human PCOLCE. | |
| 100 μg | |
| Primary | |
| Canine, Human |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PCOLCE. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| PCPE/PCPE1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5118 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only