Learn More
PCDH11Y, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PCDH11Y.
Brand: Abnova PAB17835.100ug
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation. [provided by RefSeq
Sequence: C-KYNWVTTPTTFKSpecifications
| PCDH11Y | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
| PCDH11Y | |
| PCDH11Y | |
| A synthetic peptide corresponding to amino acids at internal region of human PCDH11Y. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PCDH11Y. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| PCDH-PC/PCDH11X/PCDH22/PCDHY | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 83259 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only