Learn More
OSBP Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of OSBP.
Brand: Abnova PAB6511.100ug
Description
Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase [provided by RefSeq
Sequence: CKEKQDWSSCPDIFSpecifications
| OSBP | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| OSBP | |
| OSBP | |
| A synthetic peptide corresponding to human OSBP. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of OSBP. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| OSBP1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5007 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only