missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H3/NR1H2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.
Brand: Abnova PAB6691.100ug
This item is not returnable.
View return policy
Description
Sequence: CRLQDKKLPPLLSEISpecifications
| NR1H3/NR1H2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| LXR-a/LXRA/RLD-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10062/7376 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NR1H3 | |
| A synthetic peptide corresponding to human NR1H3, NR1H2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize an epitope corresponding to aa 429-442 of human LXR alpha protein (NP_005684) and to aa 443-456 of human LXR beta protein (NP_009052). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction