missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOA1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of NOA1.
Brand: Abnova PAB15613.100ug
This item is not returnable.
View return policy
Description
Sequence: ESDYCTAKGSEAIDRSpecifications
| NOA1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| NOA1 | |
| NOA1 | |
| A synthetic peptide corresponding to amino acids at internal region of human NOA1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of NOA1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| C4orf14/hAtNOS1/hNOA1/mAtNOS1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84273 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction