missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK7 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NEK7.
Brand: Abnova PAB18953.100ug
This item is not returnable.
View return policy
Description
NIMA-related kinases share high amino acid sequence identity with the gene product of the Aspergillus nidulans 'never in mitosis A' gene, which controls initiation of mitosis.[supplied by OMIM
Sequence: C-NTLANFRIEKKSpecifications
| NEK7 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.05-0.2 ug/mL) The optimal working dilution should be determined by the end user. | |
| NEK7 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 140609 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NEK7. | |
| In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NEK7 | |
| A synthetic peptide corresponding to amino acids at internal region of human NEK7. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction