missing translation for 'onlineSavingsMsg'
Learn More

NEIL1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of NEIL1.

Brand:  Abnova PAB6342.100ug

Product Code. 16099587

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NEIL1 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction (Bandaru et al., 2002 [PubMed 12509226]).[supplied by OMIM

Sequence: C-DIPSLEPEGTSAS
Specifications

Specifications

NEIL1
Polyclonal
Unconjugated
ELISA (1:16000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
NEIL1
NEIL1
A synthetic peptide corresponding to human NEIL1.
100 μg
Primary
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed)
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of NEIL1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
FLJ22402/FPG1/NEI1/hFPG1
Goat
Antigen affinity purification
RUO
79661
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only