Learn More
NDUFS6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NDUFS6.
Brand: Abnova PAB19021.100ug
Description
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria. The iron-sulfur protein (IP) fraction is made up of 7 subunits, including NDUFS6. See NDUFS1 (MIM 157655).[supplied by OMIM
Sequence: C-RIRFVGRQKEVNENSpecifications
| NDUFS6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| NDUFS6 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4726 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NDUFS6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NDUFS6 | |
| A synthetic peptide corresponding to amino acids at internal region of human NDUFS6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only