Learn More
NDUFS3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NDUFS3.
Brand: Abnova PAB14140.100ug
Description
This gene encodes one of the iron-sulfur protein (IP) components of mitochondrial NADH:ubiquinone oxidoreductase (complex I). Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency
Sequence: C-DTRPTVRPRNDVAHKSpecifications
| NDUFS3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NDUFS3 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4722 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NDUFS3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NDUFS3 | |
| A synthetic peptide corresponding to human NDUFS3. | |
| 100 μg | |
| Primary | |
| Approx 28 KDa band observed in human brain (frontal cortex) lysates (calculated MW of 30.2 KDa according to NP_004542.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only