missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH15, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MYH15.
Brand: Abnova PAB17850.100ug
This item is not returnable.
View return policy
Description
Sequence: TADGESLSIKEDKIQ-CSpecifications
| MYH15 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| MYH15 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 22989 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MYH15. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| MYH15 | |
| A synthetic peptide corresponding to human MYH15. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction