missing translation for 'onlineSavingsMsg'
Learn More

MTHFR Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of MTHFR.

Brand:  Abnova PAB17224.100ug

Product Code. 16042657

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Methylenetetrahydrofolate reductase (EC 1.5.1.20) catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a cosubstrate for homocysteine remethylation to methionine.[supplied by OMIM

Sequence: C-NRPTQNARETEAP
Specifications

Specifications

MTHFR
Polyclonal
Unconjugated
ELISA (1:16000) The optimal working dilution should be determined by the end user.
MTHFR
Goat
Antigen affinity purification
RUO
4524
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of MTHFR.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
MTHFR
A synthetic peptide corresponding to human MTHFR.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only