Learn More
MSC, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MSC.
Brand: Abnova PAB6020.100ug
Description
The protein encoded by this gene is a transcriptional repressor capable of binding an E-box element either as a homodimer or as a heterodimer with E2A in vitro. The encoded protein also forms heterodimers with E2A proteins in vivo. This protein is capable of inhibiting the transactivation capability of E47, an E2A protein, in mammalian cells. This gene is a downstream target of the B-cell receptor signal transduction pathway. [provided by RefSeq
Sequence: C-PLKSDWNSLDGIIRSpecifications
| MSC | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| MSC | |
| MSC | |
| A synthetic peptide corresponding to human MSC. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MSC. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ABF-1/ABF1/MYOR/bHLHa22 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9242 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only